Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.19640

Supelco

LiChrospher® Si 60 (5 µm) HPLC Columns

HPLC sorbent, CAS 7631-86-9, EC Number 231-545-4, chemical formula SiO₂.

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115709
EC Index Number:
231-545-4
NACRES:
SB.52

Product Name

LiChrospher® Si 60 (5 µm), LiChrospher® Si 60 (5 µm). CAS 7631-86-9, EC Number 231-545-4, chemical formula SiO₂.

material

stainless steel column

Agency

suitable for USP L3

product line

LiChrospher®

potency

>5000 mg/kg LD50, skin (Rabbit)

feature

endcapped: no

parameter

60 °C max. temp.

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable (Low - Medium)

surface area

350 m2/g

matrix

fully porous particle

matrix active group

Silica bonding phase

particle size

5 μm

pore size

60 Å pore size

operating pH range

2-7.5 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

200‑800 kg/m3

separation technique

hydrophilic interaction (HILIC)
normal phase

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LiChrospher® Si 60 is a versatile HPLC spherical silica sorbent with a pore diameter of 60A and a particle size of 5µm for normal phase chromatography.

Linkage

Replaces: 19640-9

Analysis Note

Specific surface area (BET): 700 - 800 m²/g
Pore volume: 0.80 - 0.90 ml/g
Pressure (Si): ≤ 30 bar
Theoretical Plates (N/m) (2-Nitrotoluene): ≥ 60000
Particle Size (d10): 4.3 - 5.1 µm
Particle Size (d50): 6.2 - 7.0 µm
Particle Size (d90): 9.0 - 9.8 µm

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service