Skip to Content
MilliporeSigma
All Photos(1)

Documents

119137

Sigma-Aldrich

CGS 21680 hydrochloride

≥96% (HPLC), solid, adenosine A2A receptor agonist, Calbiochem®

Synonym(s):

Adenosine A2A Receptor Agonist I, CGS 21680, Hydrochloride, CGS-21680, CGS21680, 3-(4-(2-((6-amino-9-((2R,3R,4S,5S)-5-(ethylcarbamoyl)-3,4-dihydroxytetrahydrofuran-2-yl)-9H-purin-2-yl)amino)ethyl)phenyl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29N7O6 · xHCl
CAS Number:
Molecular Weight:
499.52 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Adenosine A2A Receptor Agonist I, CGS 21680, Hydrochloride,

Assay

≥96% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mM

shipped in

ambient

storage temp.

−20°C

InChI

1S/C23H29N7O6/c1-2-25-21(35)18-16(33)17(34)22(36-18)30-11-27-15-19(24)28-23(29-20(15)30)26-10-9-13-5-3-12(4-6-13)7-8-14(31)32/h3-6,11,16-18,22,33-34H,2,7-10H2,1H3,(H,25,35)(H,31,32)(H3,24,26,28,29)/t16-,17+,18-,22?/m0/s1

InChI key

PAOANWZGLPPROA-OFRRTHGGSA-N

General description

CGS 21680 is a potent adenosine A2A receptors agonist (Ki = 27 nM) and xanthin derivative. CGS 21680 HCl exhibits reduced affinity for A2B receptors (Ki = 67 nM), but offers greater selectivity over A1R (Ki = 290 nM) and A3R (88.8 µM). Exerts an anti-inflammatory effect during chronic inflammation and ameliorates the tissue damage associated with collagen-induced arthritis. Been, shown to reduce CCR7 Protein Expression in THP1 macrophages. Shown to delay the progressive deterioration of motor performance and prevent a reduction in brain weight in murine model of Huntington′s disease.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a hydrochloride salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

The molecular weight of CGS 21680 Hydrochloride this compound is batch-specific due to variable water content. For batch-specific molecular weight information, please refer the vial label or the certificate of analysis. The provided molecular weight represents the baseline molecular weight without water.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cinthia Rangel-Sandoval et al.
iScience, 27(5), 109681-109681 (2024-04-29)
Pannexins are large-pore ion channels expressed throughout the mammalian brain that participate in various neuropathologies; however, their physiological roles remain obscure. Here, we report that pannexin1 channels (Panx1) can be synaptically activated under physiological recording conditions in rodent acute hippocampal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service