Skip to Content

1.18488

N-Acetyl-DL-tryptophan

greener alternative

Greener Alternative

EMPROVE® EXPERT, Ph. Eur., BP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H14N2O3
CAS Number:
Molecular Weight:
246.26
EC Index Number:
201-739-3
UNSPSC Code:
12352209
Beilstein/REAXYS Number:
89478
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

agency

BP, Ph. Eur.

product line

EMPROVE® EXPERT

form

crystalline

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

low in endotoxins

mp

204-206 °C (dec.) (lit.)

application(s)

liquid formulation
parenterals
pharma/biopharma processes

greener alternative category

storage temp.

10-30°C

SMILES string

CC(=O)NC(Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C13H14N2O3/c1-8(16)15-12(13(17)18)6-9-7-14-11-5-3-2-4-10(9)11/h2-5,7,12,14H,6H2,1H3,(H,15,16)(H,17,18)

InChI key

DZTHIGRZJZPRDV-UHFFFAOYSA-N

General description

Finding theright excipient that matches your needs as well as regulatory demands can be acomplicated challenge in formulation. With our application know-how andregulatory expertise, we support you in every step of development, scale-up,and production.As part of our Emprove® Program, our raw materials are offeredwith extensive documentation facilitating compliance of your pharma andbiopharma product, full supply chain transparency and risk mitigation. OurSAFC® portfolio of high-quality products for biopharmaceutical andpharmaceutical formulation and production withstands strict quality controlprocedures and is produced according to applicable cGMP guidelines.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Application

N-Acetyl-D,L-Tryptophanis widely used as a stabilizer in blood derived therapeutics.Due to its lowmicrobial and endotoxin limits, N-Acetyl-DL-tryptophan Emprove® Expert issuitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under N-Acetyl-DL-tryptophan


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.