Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.17945

Supelco

Potassium Test Strips, reflectometric

1.0-25.0 mg/L (K), REFLECTOQUANT®

Synonym(s):

Potassium chemical test strips, Potassium test strip for instrument reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Potassium Test, reflectometric, 1.0-25.0 mg/L (K), for use with REFLECTOQUANT

description

quantitative

Quality Level

specific analyte(s)

potassium

measuring range

1.0-25.0 mg/L (K)

compatibility

RQflex® 10 Plus
for use with REFLECTOQUANT

detection method

reflectometric

storage temp.

15-25°C

General description

Method: reflectometric 1.0 - 25.0 mg/l K Reflectoquant® plus
With this Reflectoquant test potassium can be measured as turbidity reflectometrically in drinking water, groundwater and surface water. Special applications for determination in soil are available. In alkaline solution potassium ions react with Kalignost (sodium tetraphenylborate) to form a slightly soluble precipitate. The resulting turbidity is evaluated reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to estimate the potassium concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for potassium. Further quantitative determination methods can be found with our Spectroquant tests.

Analysis Note

function control: conforms
Assay
- standard 1:
2.2 - 2.8 mg/l
- standard 2: 9.0 - 11.0 mg/l
- standard 3: 18.0 - 22.0 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service