Select a Size
About This Item
Product Name
Fluoride Test, photometric, 0.02-2.00 mg/L (F-), Spectroquant®
product line
Spectroquant®
Quality Segment
usage
sufficient for 250 tests
specific analyte(s)
fluoride
measuring range
0.02-2.00 mg/L (F-)
technique(s)
photometry: suitable
application(s)
cannabis testing
drinking water
electroplating
environmental
ground and surface water
mineral water
seawater
wastewater
compatibility
for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)
detection method
photometric (SPADNS method (As free))
storage temp.
15-25°C
General description
Fluoride ions react with part of an arsenic-free red zirconium-dye solution to form a colorless complex, which in turn results in a bleaching of the red color (SPADNS method). The concentration of the unconsumed zirconium-dye solution is subsequently determined photometrically. The method is analogous to APHA 4500-F- D.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) , which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
Legal Information
Still not finding the right product?
Explore all of our products under Fluoride Test
Signal Word
Warning
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Hazard Codes
Storage Class
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

