Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.17181

Millipore

MAS-100 Iso NT® Remote Cable

For remote operation and visual control of MAS-100 Iso line products

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

plastic cable(s)

Agency

non-compendial

feature

electronic

packaging

pkg of 1 kit

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

no temp limit

General description

Remote cable is an additional accessory designed to be specifically used for MAS-100 ISO NT®. The MAS-100 ISO NT® has been specially developed for microbial monitoring of ambient air is isolators, RABS and high-grade cleanrooms. The remote cable is not suitable for MAS-100 ISO MH® air sampler and is used for remote operation of the control unit. It has the following specifications.
  • PVC cable 4x0.5 mm2, length: 2.50 m, Diameter for mounting is 5.7 mm
  • Phoenix contact MC1,5/4-STF-3,5 Mini-Combicon
  • Push button with 2-coloured LED (red/green), diameter for mounting

Application

MAS-100 ISO NT®, remote cable is an additional accessory used for remote operation of the control unit. The MAS-100 ISO NT® sampling devices are extensively used in Pharmaceutical applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service