Skip to Content
MilliporeSigma

1152-M

Adenine

Synthetic, therefore free of any contamination with related natural products.RTECS-AU6125000Merck index number - 14, 152

Synonym(s):

Adenine, 6-Aminopurine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
UNSPSC Code:
12352208
NACRES:
NA.51
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - AU6125000
Merck USA index - 14, 152

Quality Segment

assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: 500 μg/mL

storage temp.

10-30°C

SMILES string

[nH]1cnc2ncnc2c1N

InChI

1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)

InChI key

GFFGJBXGBJISGV-UHFFFAOYSA-N

General description

Synthetic, therefore free of any contamination with related natural products.

Application


  • EFHD1 promotes osteosarcoma proliferation and drug resistance by inhibiting the opening of the mitochondrial membrane permeability transition pore (mPTP) by binding to ANT3.: This research explores the molecular mechanisms of EFHD1 in osteosarcoma, noting the involvement of adenine nucleotides in mitochondrial function regulation (Shen et al., 2024).

  • Rice Origin Tracing Technology Based on Fluorescence Spectroscopy and Stoichiometry.: This paper discusses a novel method for tracing rice origins using fluorescence spectroscopy, where adenine compounds play a crucial role in the stoichiometric analysis (Li et al., 2024).

  • Year-Long Stability of Nucleic Acid Bases in Concentrated Sulfuric Acid: Implications for the Persistence of Organic Chemistry in Venus′ Clouds.: Investigates the stability of nucleic acid bases, including adenine, in harsh chemical environments, offering insights into potential biochemical processes on Venus (Seager et al., 2024).

  • Characterization of the Mitogenome of the Genus Dendrocerus Ratzeburg (Hymenoptera: Megaspilidae) with the Specific Designed Primers.: This study characterizes the mitochondrial genome of Dendrocerus species, employing adenine-rich primers for genetic sequencing (Wang et al., 2024).

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


Still not finding the right product?

Explore all of our products under Adenine


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library