Skip to Content
MilliporeSigma

114802

AdaAhx3L3VS

A potent, covalent, cell-permeable, irreversible inhibitor of chymotrypsin-like (50-100 nM), trypsin-like (1-5 µM), and PGPH (0.5-1 µM) activities of the 20S proteasome.

Synonym(s):

AdaAhx3L3VS, Adamantane-acetyl-(6-aminohexanoyl)₃-(leucinyl)3-vinyl-(methyl)-sulfone, Proteasome Inhibitor XVI

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C50H88N6O8S
CAS Number:
Molecular Weight:
933.33
MDL number:
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥90% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, peptide vinyl sulfone that acts as a potent, irreversible inhibitor of proteasome activity. Covalently modifies the catalytically active β-subunits of the proteasome. Shown to inhibit the chymotrypsin-like (10-50 nM), trypsin-like (1-5 µM), and the peptidyl glutamyl peptide hydrolyzing (PGPH) (0.5-1 µM) activities of purified 20S proteasomes from rabbit muscle. Inhibits proteasomal degradation of deglycosylated MHC HLA-A2 heavy chain in US11+ cells (~50 µM).
A potent, covalent, cell-permeable, irreversible inhibitor of chymotrypsin-like (50-100 nM), trypsin-like (1-5 µM), and PGPH (0.5-1 µM) activities of the 20S proteasome.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
chymotrypsin-like activity of the 20S proteasome
Product does not compete with ATP.
Reversible: no
Target IC50: 50-100 nM against chymotrypsin-like activity of the 20S proteasome

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kessler, B.M., et al. 2001. Chem. Biol.8, 913.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library