Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14416

Supelco

Sulfide Test Kit, colorimetric

0.02-0.25 mg/L (S²⁻), MQuant®

Synonym(s):

Sulfide Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Sulfide Test, colorimetric, 0.02-0.25 mg/L (S2-), for use with MCOLORTEST®

specific analyte(s)

sulfide

Quality Level

measuring range

0.02-0.25 mg/L (S2-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with long test tubes 0.02-0.04-0.06-0.08-0.10-0.13-0.16-0.20-0.25 mg/l S MQuant®
The MQuant® Sulfide test (formally MColortest®) can be used for the determination of sulfide in e.g.
  • Groundwater and surface water
  • Drinking water and mineral water
  • Waters from aquaculture
  • Wastewater
  • Soils after appropriate sample pretreatment
This test is not suited for seawater.

Method
In aqueous solutions of sulfides a pH-dependent equilibrium exists between dissolved hydrogen sulfide (H2S), hydrogen sulfide ions (HS-), and sulfide ions (S2-). In the acidic range, in which practically only dissolved hydrogen sulfide is present, this reacts with dimethyl-p-phenylenediamine and iron(III) ions to form methylene blue. The sulfide concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Due to the long test tubes a sensitive measurement range can be covered.
Using the refill pack 1.18468 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Sulfide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service