Skip to Content
MilliporeSigma

1.14403

Iron Test Kit, colorimetric

0.01-0.20 mg/L (Fe), MQuant®

Synonym(s):

Iron Visual Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Iron Test, colorimetric, 0.01-0.20 mg/L (Fe), for use with MQUANT®

specific analyte(s)

iron

Quality Segment

measuring range

0.01-0.20 mg/L (Fe)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with long test tubes 0.01 - 0.02 - 0.03 - 0.04 - 0.06 - 0.08 - 0.10 - 0.15 -0.20 mg/l Fe MQuant®
The MQuant® Iron test (formally MColortest®) measures bivalent and trivalent iron in its dissolved form as well as fresh colloidal iron(III) hydroxide in
  • Groundwater and surface water, seawater
  • Drinking water and mineral water
  • Waters from aquaculture
  • Boiler and boiler feed water, cooling water
  • Industrial water
  • Wastewater and percolating water
  • Food after appropriate sample pretreatment

Method
All iron ions are reduced to iron(II) ions. In a thioglycolate-buffered medium these react with a triazine derivative to form a red-violet complex. The iron concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Due to the long test tubes a sensitive measurement range can be covered.
Using the refill pack 1.18458 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Iron Test


pictograms

CorrosionSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library