Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.12518

Millipore

Florisil® (0.150-0.250 mm)

for column chromatography

Synonym(s):

Florisil® (0.150-0.250 mm)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
215-681-1

Quality Level

pH

9.5 (20 °C, 100 g/L in H2O, slurry)

storage temp.

no temp limit

SMILES string

[Si](=O)([O-])[O-].[Mg+2]

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Analysis Note

Sieve analysis (dry sieving; >0,15 mm): ≥ 80 %
pH-value: 9.0 - 10.0

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service