Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.12169

Sigma-Aldrich

Gallium

99+

Synonym(s):

Gallium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ga
CAS Number:
Molecular Weight:
69.72
MDL number:
UNSPSC Code:
12141715
EC Index Number:
231-163-8
NACRES:
NA.22

Quality Level

form

solid

bp

2000 °C

mp

29.8 °C

density

5.904 g/cm3 at 20 °C

SMILES string

[Ga]

InChI

1S/Ga

InChI key

GYHNNYVSQQEPJS-UHFFFAOYSA-N

Analysis Note

Assay (complexometric): ≥ 99.5 %
Identity (ICP-OES): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service