Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.11931

Millipore

Casein Peptone

Encymatic digest of casein from bovine milk, suitable for biotechnology and microbiology

Synonym(s):

Peptone from casein (Tryptone), Fermtech base materials for biotechnology, Peptone from casein. Tryptone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106212
EC Index Number:
293-428-4
NACRES:
NA.74

product name

Peptone from casein (Tryptone), for biotechnology Fermtech®

biological source

bovine milk

Quality Level

Assay

12-14% amino-nitrogen basis (purity)

form

powder

manufacturer/tradename

Fermtech®

pH

6.7-7.7

solubility

800 g/L

bulk density

490 kg/m3

application(s)

microbiology

storage temp.

15-25°C

General description

Peptone from casein (Tryptone) is a pancreatic digest of casein, used as a nitrogen, vitamins, and minerals source in culture media. It supports the growth of fastidious and non-fastidious micro-organisms. It can also be used for indole production because of its high tryptophan concentration. It is used as an additive to nutrient media in the production of toxins, vaccines, enzymes, and further biological products.

Application

Peptone from casein (Tryptone) is used for detecting bacteria in dairy products, drinking and wastewater, and in antibiotic test media.

Analysis Note

Appearance: Lightly yellow to lightly brownishyellow fine powder.
Identity: passes test
Total nitrogen (N) (Kjeldahl): 12.0 - 14.0 %
Amino nitrogen (as N): 3.0 - 5.0 %
pH-value (5 %; water): 6.7 - 7.7
Sulfated ash (800 °C): ≤ 15 %
Loss on Drying (105°C): ≤ 6 %

Legal Information

FERMTECH is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service