Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
La2O3
CAS Number:
Molecular Weight:
325.81
UNSPSC Code:
12352303
EC Index Number:
215-200-5
NACRES:
NB.21
MDL number:
Quality Level
description
for inorganic trace analysis
potency
>9968 mg/kg LD50, oral (Rat)
pH
9.0 (20 °C, 50 g/L in H2O, slurry)
mp
2315 °C
density
6.51 g/cm3 at 20 °C
bulk density
1300 kg/m3
storage temp.
no temp limit
SMILES string
[La+3].[La+3].[O-2].[O-2].[O-2]
InChI
1S/2La.3O/q2*+3;3*-2
InChI key
MRELNEQAGSRDBK-UHFFFAOYSA-N
Application
- Palladium (0) nanoparticles distributed on lanthanum (III) oxide as an effective catalyst for the methanolysis of hydrazine-borane to produce hydrogen.: The study presents a novel catalyst combining Pd(0) nanoparticles with La(III) oxide for efficient hydrogen production through methanolysis of hydrazine-borane (Ruzgar et al., 2024).
- Influence of Lanthanum Precursor on the Activity of Nickel Catalysts in the Mixed-Methane Reforming Process.: This paper investigates how different lanthanum precursors affect the performance of nickel catalysts in mixed-methane reforming, highlighting La(III) oxide′s role in catalytic processes (Zakrzewski et al., 2023).
Analysis Note
Assay (complexometric, on of the ignited basis substance): ≥ 99.5 %
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.002 %
Ca (Calcium): ≤ 0.005
Cu (Copper): ≤ 0.0003 %
Fe (Iron): ≤ 0.002
Li (Lithium): ≤ 0.0001 %
Mg (Magnesium): ≤ 0.0003 %
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.002 %
Ca (Calcium): ≤ 0.005
Cu (Copper): ≤ 0.0003 %
Fe (Iron): ≤ 0.002
Li (Lithium): ≤ 0.0001 %
Mg (Magnesium): ≤ 0.0003 %
Still not finding the right product?
Explore all of our products under Lanthanum(III) oxide
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
United States Pharmacopeia Methods - Esomeprazole
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1109820025 | 04022536129266 |