Skip to Content
MilliporeSigma

1.10322

IC Multi-element standard VII

NH<SUB>4</SUB>, Ba, Ca, K, Li, Na, Mg, Mn, Sr 100 mg/L (in 1mM HNO<SUB>3</SUB>), Certipur®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Certipur®

Quality Segment

composition

NH<SUB>4</SUB>, Ba, Ca, K, Li, Na, Mg, Mn, Sr, 100 mg/L (in 1mM HNO<SUB>3</SUB>)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

multi-component solution

storage temp.

15-25°C

General description

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. Download your certificate at https://www.sigmaaldrich.com/US/en/documents-search

Analysis Note

Determination method: Ion-chromatography
Accuracy of the method: NH4, Ba, Ca, K, Li, Mg, Mn, Na, Sr; 5%
NH₄ (Ammonium): 95.0 - 105.0 mg/l
Ba (barium): 95.0 - 105.0 mg/l
Ca (calcium): 95.0 - 105.0 mg/l
K (potassium): 95.0 - 105.0 mg/l
Li (Lithium): 95.0 - 105.0 mg/l
Mg (Magnesium): 95.0 - 105.0 mg/l
Mn (Manganese): 95.0 - 105.0 mg/l
Na (sodium): 95.0 - 105.0 mg/l
Sr (Strontium): 95.0 - 105.0 mg/l

Other Notes

Visit the IC Portal to learn more

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library