Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.10230

Millipore

GSP agar

Pseudomonas Aeromonas selective agar acc. to KIELWEIN (base) for microbiology

Synonym(s):

Pseudomonas Aeromonas selective agar, Pseudomonas Aeromonas selective agar (base)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.1-7.3 (25 °C, 45 g/L in H2O, after autoclaving)

solubility

45 g/L

bulk density

820 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Pseudomonas spp.

Analysis Note

Appearance (clearness): clear - almost clear
Appearance (color): red
pH-value (25 °C): 7.0 - 7.4
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): good to very good
Growth (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): good to very good
Growth (Pseudomonas aeruginosa ATCC 10145 (WDCM 00024)): good to very good
Growth (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): good to very good
Growth (Aeromonas veronii ATCC 9071): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): none to poor
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): none
color change to red (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): +
color change to red (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): +
color change to red (Pseudomonas aeruginosa ATCC 10145 (WDCM 00024)): +
Color change to yellow (Aeromonas hydrophila ATCC 7966 (WDCM 00063)): +
Color change to yellow (Aeromonas veronii ATCC 9071): +
Incubation: 48 hours at 28 °C aerobic

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service