Skip to Content
MilliporeSigma

1.10165

Hydrochloric acid

1 M, EMPROVE® EXPERT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HCl
CAS Number:
Molecular Weight:
36.46
UNSPSC Code:
12352301
Beilstein/REAXYS Number:
1098214
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

vapor density

1.3 (vs air)

vapor pressure

3.23 psi ( 21.1 °C), 7.93 psi ( 37.7 °C)

product line

EMPROVE® EXPERT

concentration

1 M

evapn. residue

≤100 ppm

ign. residue

≤80 ppm

pH

<1 (20 °C)

bp

>100 °C (lit.)

density

1.02 g/cm3 at 20 °C, 1.18 g/mL at 20 °C

anion traces

chlorine and bromine (as Cl-): ≤1, sulfate (SO42-): ≤5 ppm, sulfite (SO32-): ≤10 ppm

cation traces

As: ≤0.1 ppm (Arsenic), Hg: ≤0.004 ppm (Mercury), Pb: ≤0.5 ppm (Heavy metals)

storage temp.

2-25°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Application

Filtered with 0.2 μm
Manufactured from Hydrochloric acid fuming 37% EMPROVE® ESSENTIAL PhEur, BP, JP, NF and purified water acc. PhEur, JP.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Hydrochloric acid


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

12 - Non Combustible Liquids



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library