Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.10107

Supelco

Arsenazo III

metal indicator

Synonym(s):

Arsenazo III, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthaline-3,6-disulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18As2N4O14S2
CAS Number:
Molecular Weight:
776.37
MDL number:
UNSPSC Code:
12352103
EC Index Number:
216-788-6
NACRES:
NA.21

product name

Arsenazo III, metal indicator

Quality Level

form

solid

loss

≤10% loss on drying, 110°C

pH

2.3 (20 °C, 10 g/L in H2O)

bulk density

850 kg/m3

λmax

569-576 nm (buffer pH 10.0)

storage temp.

2-30°C

InChI

1S/C22H18As2N4O14S2/c29-21-18-11(9-16(43(37,38)39)19(21)27-25-14-7-3-1-5-12(14)23(31,32)33)10-17(44(40,41)42)20(22(18)30)28-26-15-8-4-2-6-13(15)24(34,35)36/h1-10,29-30H,(H2,31,32,33)(H2,34,35,36)(H,37,38,39)(H,40,41,42)

InChI key

UQHVTNUJRKELCE-UHFFFAOYSA-N

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (buffer pH 10.0): 569 - 576 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.025 g/l; buffer pH 10.0; calculated on dried substance): 300 - 450
Loss on drying (110 °C): ≤ 10 %
Suitability as indicator: passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service