Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09906

Supelco

Silver standard

1000 mg Ag, (AgNO₃ in HNO₃ 5%) Titrisol®

Synonym(s):

Silver Calibration Solution for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg Ag in 5% HNO3

technique(s)

AAS: suitable
ICP: suitable

pH

0.5 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Application


  • Alternative electrolytes in breath metabolomics: Research on untargeted breath metabolomics employing secondary-electrospray ionization highlighted the use of silver-based electrolyte solutions, underscoring their importance in improving detection sensitivity and analytical reliability in biochemical assays (Wuthrich, Zenobi, and Giannoukos, 2024).

  • Biogenic synthesis of silver nanoparticles: A study investigating silver nanoparticles synthesized by Salvia species illustrated their multifunctional properties, including catalysis, sensing, antimicrobial, and antioxidant capabilities, relevant for scientific laboratory assays and biochemistry applications (Ihsan et al., 2024).

Analysis Note

content (Ag): 1000 mg / Ampoule
Determination method: Argentometric titration.
(traceable to NIST - SRM 999a)
Accuracy of the method: +/- 2 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service