Skip to Content
MilliporeSigma

1.09883

Buffer concentrate

(citrate/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 3.00 ± 0.02 (20°C) Titrisol®

Synonym(s):

Buffer concentrate for 500 ml buffer solution, pH 3.00 ± 0.02 (20°C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12161704
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

agency

according to DIN 19266

product line

Titrisol®

pH

3.00 (20 °C in H2O)

density

1.06 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The Titrisol® reference buffer concentrate [citrate/hydrochloric acid; pH=3.00 ± 0.02 (20°C)] is analyzed in the ISO/IEC 17025 accredited calibration laboratory with a combined glass electrode and 5-point calibration according to DIN 19268 against a certified primary standard reference material following DIN 19266.

The Titrisol® concentrated buffer material combines ease of use and maximum precision for the routine calibration and monitoring of pH instruments. To prepare the ready-to-use pH buffer solution, simply dilute the Titrisol® buffer concentrate.

Download your certificate at https://www.sigmaaldrich.com/US/en/documents-search  to view certified values, including uncertainty and expiry date.

Features and Benefits

  • Traceable to primary standard reference materials from NIST and PTB
  • Analytical standard Grade
  • Uncertainty data and traceability are documented in a comprehensive and batch-specific Certificate of Analysis.

Analysis Note

pH-value (20 °C): 3.00 ± 0.02
Measured after dilution to 500 ml with a combination glass electrode after multiple point calibration (according to DIN 19268) with reference materials for the preparation of standard buffer solutions according to DIN 19266.

Other Notes

Please visit Physical & Chemical Property Standards to explore a wide selection of standards and certified reference materials (CRMs) for many tests parameters.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Buffer concentrate


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library