Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09875

Millipore

Bacillus cereus Selective Supplement

selective by defined nutrients for Bacillus cereus, pkg of 10 vials, for the preparation of MYP Agar Base

Synonym(s):

Bacillus Cereus, MYP Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Index Number:
215-774-7
NACRES:
NA.74

description

GranuCult™ Mannitol egg yolk polymyxin agar (base) (1.05267)

Quality Level

composition

Polymyxin B sulfate, 50000 (IU)

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

selective by defined nutrients for Bacillus cereus

SMILES string

[S](=O)(=O)(O)O.N1CCC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C1=O)CCO)CCN)CCN)CC(C)C)Cc2ccccc2)CCN)NC(=O)C(NC(=O)C(NC(=O)C(NC=O)CCN)CCO)CCN

InChI

1S/C48H82N16O13.H2O4S/c1-27(2)24-37-47(76)59-32(11-19-52)41(70)56-31(10-18-51)43(72)61-35(14-22-65)39(68)54-21-13-34(45(74)57-33(12-20-53)44(73)64-38(48(77)63-37)25-28-6-4-3-5-7-28)60-42(71)30(9-17-50)58-46(75)36(15-23-66)62-40(69)29(8-16-49)55-26-67;1-5(2,3)4/h3-7,26-27,29-38,65-66H,8-25,49-53H2,1-2H3,(H,54,68)(H,55,67)(H,56,70)(H,57,74)(H,58,75)(H,59,76)(H,60,71)(H,61,72)(H,62,69)(H,63,77)(H,64,73);(H2,1,2,3,4)

InChI key

HNDFYNOVSOOGDU-UHFFFAOYSA-N

General description

Bacillus cereus Selective Supplement contains Polymyxin B sulfate in lyophilized form. It suppresses the growth of accompanying bacterial flora during culturing Bacillus cereus.

Application

Bacillus cereus Selective Supplement is an additive for the preparation of MYP Agar Base for the enumeration of Bacillus cereus from food and animal feed.

Linkage

Replaces: 1.09875

Analysis Note

Growth promotion test in accordance to ISO 11133.
Inoculum on reference medium
- Bacillus cereus ATCC 11778 (WDCM 00001):
10 - 100
- Bacillus subtilis ATCC 6633 (WDCM 00003): 10 - 100
Recovery on test medium
- Bacillus cereus ATCC 11778 (WDCM 00001):
≥ 70 %
- Bacillus subtilis ATCC 6633 (WDCM 00003): no limit
Colony colour
- Bacillus cereus ATCC 11778 (WDCM 00001):
pink
- Bacillus subtilis ATCC 6633 (WDCM 00003): yellow
Precipitate
- Bacillus cereus ATCC 11778 (WDCM 00001):
+
- Bacillus subtilis ATCC 6633 (WDCM 00003): -
Growth
- Escherichia coli ATCC 25922 (WDCM 00013):
no growth
- Escherichia coli ATCC 8739 (WDCM 00012): no growth
Incubation: Bacillus 24-48 hrs. 30°c aerobic; other strains 48 hrs. 30°C

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Interview: Accurate microbiology QC methods for infant formula testing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service