Skip to Content
MilliporeSigma

1.09708

Ethyl methyl ketone

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Ethyl methyl ketone, 2-Butanone, Methyl ethyl ketone, MEK

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
UNSPSC Code:
12191502
EC Index Number:
201-159-0
NACRES:
NA.05
MDL number:
Assay:
≥99.5% (GC)
Grade:
ACS reagent
Bp:
79.6 °C/1013 hPa
Vapor pressure:
105 hPa ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

vapor pressure

105 hPa ( 20 °C)

product line

EMSURE®

assay

≥99.5% (GC)

form

liquid

autoignition temp.

514 °C

potency

3400 mg/kg LD50, oral (Rat), >8000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-11.5 % (v/v)

dilution

(for analytical testing)

impurities

≤0.0002 meq/g Alkalinity, ≤0.0003% Substances reducing potassium permanganate (as O), ≤0.0005 meq/g Titrable acid, ≤0.05% Acetone (GC), ≤0.05% Methanol (GC), ≤0.05% Water, ≤0.1% 2-Butanol (GC)

evapn. residue

≤0.001%

color

clearAPHA: ≤10

pH

7 (20 °C)

bp

79.6 °C/1013 hPa

mp

-86 °C

transition temp

flash point -4 °C

solubility

292 g/L

density

0.805 g/cm3 at 20 °C

cation traces

Al: ≤0.00002%, B: ≤0.000002%, Ba: ≤0.00001%, Ca: ≤0.00005%, Cd: ≤0.000005%, Co: ≤0.000002%, Cr: ≤0.000002%, Cu: ≤0.000002%, Fe: ≤0.00001%, Mg: ≤0.000005%, Mn: ≤0.000002%, Ni: ≤0.000002%, Pb: ≤0.00001%, Sn: ≤0.00001%, Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

O=C(CC)C

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • In Situ Synthesis of Environmentally Friendly Waterborne Polyurethane Extended with Regenerated Cellulose Nanoparticles for Enhanced Mechanical Performances: Ethyl methyl ketone is utilized in the synthesis process of eco-friendly polyurethane materials, contributing to the development of sustainable composite materials with improved mechanical properties (Choi et al., 2023).

  • Comparative Studies on the Physicochemical and Volatile Flavour Properties of Traditional Deep Fried and Circulating-Air Fried Hairtail (Trichiurus lepturus): Ethyl methyl ketone′s solvent properties are utilized in the analysis of food flavors, particularly in comparing cooking methods and their impact on flavor profile, which is vital for food science research (Ding et al., 2022).

Other Notes

Discover EMSURE - to be more than sure!
Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Ethyl methyl ketone


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

30.2 °F - closed cup

flash_point_c

-1 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library