Skip to Content
MilliporeSigma

1.09532

pH-indicator strips pH 7.5 - 14

non-bleeding, colorimetric, pH range 7.5-14.0, graduations and accuracy: 0.5 pH unit, MQuant®

Synonym(s):

pH dip stick pH 7.5 - 14, pH test strips pH 7.5 - 14

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41113034
NACRES:
NB.35
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

pH-indicator strips pH 7.5 - 14, non-bleeding, colorimetric, pH range 7.5-14.0, graduations and accuracy accuracy: 0.5 pH unit, MQuant®

product line

MQuant®

Quality Segment

feature

non-bleeding

graduations and accuracy measuring range

accuracy: 0.5 pH unit

pH range

7.5-14.0

application(s)

agriculture
beverage testing
disinfection control
food testing
wastewater
water monitoring

detection method

colorimetric

storage temp.

2-30°C

General description

The MQuant® pH tests are suited for all media in environmental analysis and in industrial in-process controls. Our special method of manufacture gives the pH indicator strips decisive advantages over the indicator paper: since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it. Using these test strips is fairly easy, the pH value is measured semiquantitatively by visual comparison of the three reaction zones on the test strip with the fields of the color scale, perfectly matching the reaction color. The clever Safetyedge boxes allow for safe storage and convenient withdrawal of the test strips. FFor tests for other parameters that require a specific pH range of the sample (e.g. SpectroQuant®, see instructions of use), we recommend the use of MQuant® pH test strips as pre-test.
non-bleeding pH 7.5 - 8.0 - 8.5 - 9.0 - 9.5 - 10.0 - 10.5 - 11.0 - 11.5 - 12.0 - 12.5 - 13.0 - 14.0 MQuant®

Other Notes

Replaces: 9587-1; 9587-3; 9587

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library