Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09478

Supelco

Ammonium buffer solution

for complexometry (ammonium chloride/ammonia) pH = 10-11

Synonym(s):

Ammonium Salt Buffer Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161705
NACRES:
NA.21

Quality Level

form

liquid

technique(s)

titration: suitable

pH

10.5 (20 °C in H2O)

density

0.978 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Ultratrace Detection of Nickel(II) Ions in Water Samples Using Dimethylglyoxime-Doped GQDs as the Induced Metal Complex Nanoparticles by a Resonance Light Scattering Sensor.: This study employs ammonium buffer solution in a novel sensor setup for the ultra-sensitive detection of nickel ions, highlighting its potential for environmental monitoring and pollution control (Pimsin N et al., 2021).

  • Simple flow injection colorimetric system for determination of paraquat in natural water.: Ammonium buffer solution is utilized in a flow injection analysis system, simplifying the detection of paraquat in water, which can be crucial for ensuring water safety and environmental protection (Chuntib P et al., 2015).

  • A simple and sensitive flow-injection on-line preconcentration coupled with hydride generation atomic fluorescence spectrometry for the determination of ultra-trace lead in water, wine, and rice.: This method uses ammonium buffer solution to enhance the sensitivity and selectivity of lead detection, important for food safety and public health (Wu H et al., 2007).

  • On-line organoselenium interference removal for inorganic selenium species by flow injection coprecipitation preconcentration coupled with hydride generation atomic fluorescence spectrometry.: Utilizes ammonium buffer to segregate and quantify selenium in complex samples, a critical advancement for environmental and health-related studies (Wu H et al., 2007).

  • On-line arsenic co-precipitation on ethyl vinyl acetate turning-packed mini-column followed by hydride generation-ICP OES determination.: In this research, ammonium buffer solution plays a crucial role in the online preconcentration technique for arsenic, essential for monitoring arsenic contamination in various matrices (Gil RA et al., 2007).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service