Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09440

Millipore

MAS-100 Iso NT® Tri-Clamp,

stainless steel, To facilitate mounting of the sampling head

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

stainless steel

Agency

non-compendial

feature

installed

packaging

pkg of 3 units

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100® Iso

shipped in

ambient

storage temp.

no temp limit

General description

MAS-100 ISO NT® Tri-Clamp is an additional accessory for MAS-100 ISO NT® and MAS-100 ISO MH® air samplers. MAS 100 Iso line is designed to monitor air microbiology in isolators and RABS. Like all other MAS-100® air samplers, it′s based on the Andersen impaction principle. All electronic and moving parts are located outside the critical zone, whilst one stainless steel sampling head for standard 90 to 100 mm Petri dishes collects samples in the containment zone. A Stainless-steel tri-clamp of 21.7mm internal diameter and these fittings can be used to facilitate mounting and removing the sampling head. The silicone gasket is used to seal the tri clamp connections on instruments of the MAS-100 Iso product family.

Application

MAS-100 ISO NT® Tri-Clamp Stainless Steel is an additional accessory used to facilitate mounting and removing the sampling head of the MAS-100 ISO NT® and MAS-100 ISO MH®. Microbial air sampling devices are extensively used in Pharmaceutical industries.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service