Skip to Content
MilliporeSigma

1.09014

Hoechst wax C micropowder

tabletting aid for X-ray fluorescence analysis

Synonym(s):

Hoechst wax C micropowder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H76N2O2
CAS Number:
Molecular Weight:
593.02
UNSPSC Code:
12352402
EC Index Number:
203-755-6
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.000023 Pa ( 20 °C)

Quality Segment

form

solid

potency

>20000 mg/kg LD50, oral (Rat)

mp

144 °C

transition temp

flash point 270 °C (DIN 51758)

density

1 g/cm3 at 20 °C

bulk density

210 kg/m3

storage temp.

15-25°C

SMILES string

O=C(NCCNC(=O)CCCCCCCCCCCCCCCCC)CCCCCCCCCCCCCCCCC

InChI

1S/C38H76N2O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-37(41)39-35-36-40-38(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-36H2,1-2H3,(H,39,41)(H,40,42)

InChI key

RKISUIUJZGSLEV-UHFFFAOYSA-N

Analysis Note

Bi (Bismuth): ≤ 10 µg/g
Co (Cobalt): ≤ 5 µg/g
Cr (Chromium): ≤ 5 µg/g
Cu (Copper): ≤ 5 µg/g
Fe (Iron): ≤ 5 µg/g
Mn (Manganese): ≤ 5 µg/g
Ni (Nickel): ≤ 5 µg/g
Pb (Lead): ≤ 10 µg/g
Sr (Strontium): ≤ 5 µg/g
Tl (Thallium): ≤ 10 µg/g
Zn (Zinc): ≤ 5 µg/g


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

518.0 °F - DIN 51758

flash_point_c

270 °C - DIN 51758



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library