Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.08633

Supelco

Xylenes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12191501
NACRES:
NA.21

grade

ACS reagent

Quality Level

form

liquid

mp

-47 °C

transition temp

flash point 26 °C

General description

isomeric mixture for analysis Emparta® ACS

Storage and Stability

Store at +5°C to +30°C.

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service