Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.08495

Millipore

L-Valine

for biochemistry

Synonym(s):

L-Valine, Val

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-773-6
NACRES:
NA.21

Quality Level

form

solid

pH

5.5-6.5 (20 °C, 100 g/L in H2O)

mp

315 °C

solubility

85 g/L

density

1.23 g/cm3 at 20 °C

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

NC(C(C)C)C(=O)O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)

InChI key

KZSNJWFQEVHDMF-UHFFFAOYSA-N

General description

for biochemistry

Application

L-Valine for biochemistry. CAS 72-18-4, pH 5.5 - 6.5 (100 g/l, H₂O, 20 °C).

Analysis Note

Assay (perchloric acid titration)≥ 99.0 %Identity (IR-spectrum)passes testAppearancewhite, crystalline powder or colorless crystalsAppearance of solution (50 g/l; water)clear and colorlessSpec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l)+23.8 to +24.8Heavy metals (as Pb)≤ 0.001 %NH₄ (Ammonium)≤ 0.01 %Foreign amino acids≤ 0.5 %Other ninhydrine positive substances≤ 0.1 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service