Skip to Content
MilliporeSigma

1.08462

Uranine AP (C.I. 45350)

concentrated for the examination of subterranean waters

Synonym(s):

Uranine AP (C.I. 45350)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
UNSPSC Code:
12171501
EC Index Number:
208-253-0
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

potency

6720 mg/kg LD50, oral (Rat)

technique(s)

titration: suitable

loss

≤7% loss on drying, 110°C

color

red-brown

pH

8.3 (20 °C, 10 g/L in H2O)

mp

>300 °C

solubility

500 g/L

bulk density

600 kg/m3

λmax

489-491 nm (buffer pH 8.0)

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]c1cc2c(cc1)C(=C4C(=CC(=O)C=C4)O2)c3c(cccc3)C(=O)[O-]

InChI

1S/C20H12O5.2Na/c21-11-5-7-15-17(9-11)25-18-10-12(22)6-8-16(18)19(15)13-3-1-2-4-14(13)20(23)24;;/h1-10,21H,(H,23,24);;/q;2*+1/p-2

InChI key

NJDNXYGOVLYJHP-UHFFFAOYSA-L

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Appearance: Reddish-brown powder, eventually with granular and bright parts.
Absorption maximum λmax. (buffer pH 8.0): 489 - 491 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; buffer pH 8.0; calc. on dried substance): ≥ 1800
Loss on drying (110 °C): ≤ 7 %


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

423.7 °F - Pensky-Martens closed cup

flash_point_c

217.6 °C - Pensky-Martens closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library