Skip to Content
MilliporeSigma

1.08312

Total Hardness Test Kit, titrimetric

titrimetric, for use with MCOLORTEST®

Synonym(s):

Total Hardness Visual Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Total Hardness Test, titrimetric, for use with MCOLORTEST®

specific analyte(s)

calcium, magnesium

Quality Segment

compatibility

for use with MCOLORTEST®

detection method

titrimetric

storage temp.

15-25°C

General description

Method: titrimetric with dropping bottle MColortest®
The test measures the content of salts of the alkaline earth metals calcium, magnesium, strontium and barium in groundwater and surface water, drinking water, mineral water and boiler water.
This MQuant® Total Hardness test (formally MColortest®) is suitable for the titrimetric determination using a convenient dropping bottle. It can be used in:
  • Groundwater, surface water, and seawater
  • Waters from aquaculture
  • Drinking water and mineral water
  • Swimming-pool water
  • Boiler water
  • Industrial water

Method
Calcium and magnesium ions react with an indicator to form a red complex compound. The indicator is released from this compound by titration with a solution of ethylenedinitrilotetraacetic acid disodium salt dihydrate (Titriplex® III). At the titration end-point the color changes to green. The total hardness is determined from the consumption of titration solution.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Total Hardness Test


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library