Skip to Content
MilliporeSigma

1.08191

Fluid Thioglycollate medium

GranuCult® prime, APHA, FDA-BAM Medium M146, granular, for anaerobic bacteria, pkg of 5 kg, pkg of 500 g

Synonym(s):

FTM, Fluid thioglycolic medium, FTG medium,

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213
Manufacturer/tradename:
GranuCult® prime
Agency:
APHA, FDA-BAM Medium M146
Ph:
6.9-7.3 (25 °C, 30 g/L in H2O, after autoclaving)
Form:
granular
Suitability:
anaerobic bacteria
Application(s):
microbiology
pharmaceutical
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

APHA, FDA-BAM Medium M146

sterility

non-sterile

form

granular

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 5 kg, pkg of 500 g

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

6.9-7.3 (25 °C, 30 g/L in H2O, after autoclaving)

solubility

30 g/L

bulk density

600 kg/m3

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

anaerobic bacteria

General description

Fluid Thioglycollate Medium (FTG medium) is a culture medium used for the detection and growth of various microorganisms, including anaerobic and aerobic bacteria. The medium contains a variety of nutrients, including tryptic soy broth or Casein hydrolysate, yeast extract, glucose, and sodium chloride, as well as a reducing agent such as thioglycollate, which helps to create an oxygen gradient that can facilitate the growth of anaerobic bacteria. FTG medium also contains a redox indicator called resazurin, which changes from pink to red in the presence of oxygen.

Application

FTG medium is used in various applications, including sterility testing of biopharmaceutical products, antimicrobial susceptibility testing, environmental monitoring, and quality control purposes in the laboratory setting. The medium can detect both aerobic and anaerobic microorganisms and is a versatile tool for diverse microbiology applications.

Features and Benefits

Our Granulated media are:
  • Safer - Significant reduction of inhalation of toxic and allergenic substances resulting in fewer allergic responses. Less contamination of working tools and environment
  • Accurate - No separation of components and no clumping, even under humid or warm conditions, ensure a uniform product with higher performance accuracy
  • Fast - Rapid and uniform dissolution in water
  • Easier - Easier handling and weighing due to the better flow properties. Media do not stick to the flask or container
  • Reliable - Homogenous distribution of ingredients guarantees high reproducibility even if only small amounts of media are used.

Other Notes

Replaces: 1.08191.5007

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.


Still not finding the right product?

Explore all of our products under Fluid Thioglycollate medium


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library