Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.07712

Supelco

Sea sand

purified by acid and calcined for analysis

Synonym(s):

Sea sand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
11111701
EC Index Number:
238-878-4
NACRES:
NA.21

Quality Level

form

solid

impurities

≤0.2% HCl solubles

loss

≤0.05% loss on ignition, 900 °C

particle size

0.1-0.315 mm (>=85%)

pH

5-8 (20 °C, 400 g/L in H2O, slurry)

bp

>2200 °C/1013 hPa

mp

1713 °C

density

2.65 g/cm3 at 20 °C

bulk density

1400 kg/m3

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤0.01% (acid soluble)

storage temp.

no temp limit

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Analysis Note

Substances soluble in hydrochloric acid: ≤ 0.2 %


Chloride (Cl): ≤ 0.005 %


Acid-soluble Iron (Fe): ≤ 0.01 %


Loss on ignition (900 °C): ≤ 0.05 %


Particle size (0.1 - 0.315 mm): ≥ 85%

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service