Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07687

Millipore

Sucrose

for biochem. Reag. Ph Eur

Synonym(s):

Sucrose, Saccharose, α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-334-9
NACRES:
NA.79
Pricing and availability is not currently available.

Agency

reag. Ph. Eur.

Quality Level

form

solid

potency

29700 mg/kg LD50, oral (Rat)

pH

7 (20 °C, 100 g/L in H2O)

mp

185-187 °C (lit.)

bulk density

800‑950 kg/m3

storage temp.

15-25°C

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for biochem. Reag. Ph Eur

Application

Sucrose (saccharose) for biochem. Reag. Ph Eur. CAS 57-50-1, pH 7 (100 g/l, H₂O, 20 °C).

Analysis Note

Identitypasses testAppearance of solution (50%; water)passes testColor number≤ 45Spec. rotation (α 20/D; 26%; water)66.3 - 67.0 °Electrical conductivity≤ 35 µS/cmSulfite (as SO₂)≤ 10 ppmDextrinspasses testReducing sugarspasses testResidual solvents (ICH Q3C)excluded by production processLoss on Drying (105 °C)≤ 0.1%Endotoxins<250

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service