Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07144

Millipore

Papain

(from Carica papaya) water-soluble 30000 USP-U/mg for biochemistry EC 3.4.22.2

Synonym(s):

Papain Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

form

solid

pH

4-7 (25 °C, 1 g/L in H2O)

bulk density

800 kg/m3

storage temp.

2-30°C

Application


  • Novel biotechnological formulations of cysteine proteases, immobilized on chitosan. This study focuses on the development and characterization of innovative biotechnological formulations of cysteine proteases including papain, which are immobilized on chitosan to enhance their stability and activity. Such immobilization has significant implications for biopharmaceutical manufacturing processes, offering potential advancements in the stability and efficiency of biotechnological applications (Holyavka et al., 2021).

  • A novel aspartic protease from Rhizomucor miehei expressed in Pichia pastoris and its application on meat tenderization and preparation of turtle peptides. While primarily focusing on a novel aspartic protease, this research discusses comparative proteolytic activities including those of papain, highlighting its utility in improving the tenderness and quality of meat products, thus supporting its industrial application in food processing and life science manufacturing (Sun et al., 2018).

  • Comparison of the proteolytic activities of new commercially available bacterial and fungal proteases toward meat proteins. This comparative study assesses the efficacy of various proteases including papain in the breakdown of meat proteins. Papain′s effective proteolytic action is beneficial for its use in industrial and research settings, particularly in food science applications where efficient protein digestion is critical (Ha et al., 2013).


Analysis Note

Appearance (colour): yellowish-brown
Appearance (description): fine powder
Activity (Casein, pH 6.0, 40 °C): ≥ 30000 USP-U/mg
USP reference standard: calculated on USP reference standardJ0H138 (activity:6800 USP-U/mg)

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I interest to your papain product. However, in the past we used papain in TU/mg unit. could you please inform me about how to convert TU/mg to USP unit. Thank you.

    1 answer
    1. 5047- USP U/mg = 90T U/mg, therefore 30000 USP-U/mg = 534.9790TU/mg.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service