Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07037

Supelco

Cyclohexane

for analysis EMPARTA® ACS

Synonym(s):

Cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
MDL number:
UNSPSC Code:
12191502
grade:
ACS reagent
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

grade

ACS reagent

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

autoignition temp.

500 °F

expl. lim.

9 %

dilution

(for routine analytical applications)

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

storage temp.

20-25°C

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Phytochemical Profiling and Biological Potential of Prunus dulcis Shell Extracts.: Cyclohexane is utilized here for the extraction and analysis of bioactive compounds in almond shells, illustrating its application in natural product chemistry and analytical methodologies. The study underscores cyclohexane′s effectiveness in extracting valuable phytochemicals for potential therapeutic and commercial use (Ben Khadher T et al., 2023).

  • Determining the Antibacterial Effect of Mentha Longifolia Essential Oil on Cariogenic Bacteria and Its Compounds: an in vitro Study.: In this study, cyclohexane is employed as a solvent to isolate essential oils for antibacterial testing, highlighting its critical role in enhancing the efficacy of natural antimicrobial agents. The research contributes to the development of cyclohexane-based methods for studying and applying essential oils in dental health (Shazdehahmadi F et al., 2023).

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service