Skip to Content
MilliporeSigma

1.06906

L-Ornithine monohydrochloride

for biochemistry

Synonym(s):

L-Ornithine monohydrochloride, L(+)-2,5-Diamino valeric acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
168.62
EC Index Number:
221-678-6
UNSPSC Code:
12352209
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

potency

10000 mg/kg LD50, oral (rat)

mp

245 °C

solubility

100 g/L

bulk density

470 kg/m3

storage temp.

2-30°C

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H

InChI key

GGTYBZJRPHEQDG-UHFFFAOYSA-N

General description

for biochemistry

Application

L-Ornithine monohydrochloride for biochemistry. CAS 3184-13-2, molar mass 168.62 g/mol.

Analysis Note

Assay (perchloric acid titration, calculated on dried substance)99.0 - 102.0 %Identity (IR-spectrum)passes testAppearancewhite, crystalline powder or colorless crystalsAppearance of solution (25 g/l; CO₂-free water)clear and not more intense in color than reference solution B9Spec. rotation (α 20/D, 40 g/l, hydrochloric acid 6 mol/l, calc. on dried substance)+23,0 to +25,0Sulfate (SO₄)≤ 200 ppmHeavy metals (as Pb)≤ 10 ppmAs (Arsenic)≤ 5 ppmCa (Calcium)≤ 100 ppmCo (Cobalt)≤ 5 ppmFe (Iron)≤ 5 ppmK (Potassium)≤ 20 ppmMg (Magnesium)≤ 10 ppmNa (Sodium)≤ 50 ppmZn (Zinc)≤ 5 ppmNinhydrin-positive substances (LC)(any ninhydrin-positive impurity)≤ 0.2 %Ninhydrin-positive substances (LC) (ammonium (570 nm))≤ 0.02 %Ninhydrin-positive substances (LC)(total impurities)≤ 1.0 %Loss on drying (105 °C)≤ 0.5 %Other ninhydrine positive substances≤ 0.1 %Loss on drying (105 °C; 3 h)≤ 0.3 %


Still not finding the right product?

Explore all of our products under L-Ornithine monohydrochloride


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
106906901004022536692784
106906100004022536084466
106906002504022536084442
106906010004022536084459