Skip to Content
MilliporeSigma

1.06900

Oil bath filling

for oil baths up to about 250°C

Synonym(s):

Oil bath filling

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41103715
EC Index Number:
232-315-6
NACRES:
NB.51
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<0.1 hPa ( 20 °C)

Quality Segment

form

liquid

potency

>5000 mg/kg LD50, oral (Rat), >3600 mg/kg LD50, skin (Rabbit)

expl. lim.

0.6-6.5 % (v/v)

matrix

Paraffin wax base material

viscosity

~5000 mPa.s(20 °C)

kinematic viscosity

965 cSt(40 °C)

bp

>316 °C/1013 hPa

transition temp

flash point 318 °C

density

0.94 g/cm3 at 15 °C

storage temp.

2-30°C

General description

Silica gel is a highly porous textured matrix that is generally used as a drying agent to dry all gases and liquids.

Application

Oil bath filling is used for oil baths with a temperature range of around 250°C. These oil baths are ideally used for all temperature control applications.
Silica gel can be used in desiccators; protecting moisture-sensitive substances during storage and transport; maintaining the dryness of anhydrous solvents; packing drying towers to dry gases and solvents. Application temperature capacity is practically independent of temperature up to approx. 65 °C, but decreases significantly at higher temperatures.

Analysis Note

Density (d 20 °C/ 4 °C): 0.92 - 0.96
Flash point: ~300
Viscosity (mPa s, 20°C): ~5000
Suitability for oil baths: passes test


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

604.4 °F

flash_point_c

318 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library