Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.06648

Supelco

Sodium sulfate decahydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Sodium sulfate decahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O4S · 10H2O
CAS Number:
Molecular Weight:
322.20
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-820-9
NACRES:
NA.21
grade:
ACS reagent
form:
crystals

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (alkalimetric)
98.5-101% dry basis (alkalimetric)

form

crystals

impurities

≤0.01% Insoluble matter

loss

52.0-57.0% loss on drying, 130 °C

pH

5.2-8.0 (25 °C, 50 g/L in H2O, anhydrous substance)

mp

32.4 °C

density

1.464 g/cm3

bulk density

700 kg/m3

anion traces

chloride (Cl-): ≤0.0003%
phosphate (PO43-): ≤0.0005%

cation traces

As: ≤0.0001%
Ca: ≤0.002%
Fe: ≤0.0005%
K: ≤0.002%
Mg: ≤0.001%
N: ≤0.0003%
heavy metals (as Pb): ≤0.0003%

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O

InChI

1S/2Na.H2O4S.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2

InChI key

RSIJVJUOQBWMIM-UHFFFAOYSA-L

Related Categories

Application


  • Nanoscale Stabilization Mechanism of Sodium Sulfate Decahydrate at Polyelectrolyte Interfaces: Discusses the stabilization mechanisms at the nanoscale, providing insights into sodium sulfate decahydrate′s applications in polyelectrolyte-enhanced processes, relevant in materials science and engineering (Akamo et al., 2024).

Analysis Note

Assay (alkalimetric): ≥ 99.0 %
Assay (alkalimetric, calculated on dried substance): 98.5 - 101.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water; 25 °C): 5.2 - 8.0
Acidity or alkalinity: passes test
Chloride (Cl): ≤ 0.0003 %
Phosphate (PO₄): ≤ 0.0005 %
Total nitrogen (N): ≤ 0.0003 %
Heavy metals (as Pb): ≤ 0.0003 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.002 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.002 %
Mg (Magnesium): ≤ 0.001 %
Loss on drying (130 °C): 52.0 - 57.0 %
corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service