Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.06597

Supelco

Sodium metaperiodate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Sodium metaperiodate, Sodium periodate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
INaO4
CAS Number:
Molecular Weight:
213.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-197-6
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (iodometric)
99.8-100.3% dry basis

form

crystals

pH

5.2 (20 °C, 50 g/L in H2O)

mp

270 °C (decomposes)

density

3.87 g/cm3 at 20 °C

bulk density

2900 kg/m3

anion traces

other halogens (as Cl-): ≤0.01%
sulfate (SO42-): ≤0.005%

cation traces

Mn: ≤0.0001%

storage temp.

2-30°C

InChI

1S/HIO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

JQWHASGSAFIOCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Use of a thermal lens microscope in integrated catecholamine determination on a microchip.: Sodium metaperiodate was used in this study for the determination of catecholamines using a microchip, showcasing its application in advanced analytical techniques (Sorouraddin et al., 2001).

  • Ultrastructural staining with sodium metaperiodate and sodium borohydride.: This research employs sodium metaperiodate for ultrastructural staining, enhancing visualization techniques in histochemistry (Lobo et al., 2002).

  • Dextran functionalized surfaces via reductive amination: morphology, wetting, and adhesion.: Sodium metaperiodate was used to functionalize dextran surfaces, improving the properties of biomaterials for various applications (Miksa et al., 2006).

  • Assay of labile estrogen o-quinones, potent carcinogenic molecular species, by high performance liquid chromatography-electrospray ionization tandem mass spectrometry with phenazine derivatization.: This study utilized sodium metaperiodate in the assay of labile estrogen o-quinones, advancing analytical chemistry techniques for detecting carcinogenic compounds (Yamashita et al., 2010).

  • One-step immobilization of antibodies for α-1-fetoprotein immunosensor based on dialdehyde cellulose/ionic liquid composite.: Sodium metaperiodate was key in the one-step immobilization of antibodies on a dialdehyde cellulose/ionic liquid composite, advancing immunosensor technology for detecting α-1-fetoprotein (Shen et al., 2015).

Linkage

Replaces: SX0695-5; SX0695-5

Analysis Note

Assay (iodometric): ≥ 99.0 %
Assay (out of dried substance): 99.8 - 100.3 %
Identity: passes test
Sulfate (SO₄): ≤ 0.005 %
other halogens (as Cl): ≤ 0.01 %
Mn (Manganese): ≤ 0.0001 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1C - STOT RE 1

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service