Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.06467

Supelco

Sodium hydroxide

granulated EMPLURA®

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
EC Index Number:
215-185-5
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

>1 (vs air)

Quality Level

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

product line

EMPLURA®

Assay

≥99% NaOH basis (acidimetric)

form

granular

pH

>14 (20 °C, 100 g/L in H2O)

mp

318 °C (lit.)

solubility

1090 g/L

density

2.13 g/mL at 20 °C

anion traces

carbonate (as Na2CO3): ≤1%
chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.002%
Fe: ≤0.002%
heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Silver Nanoparticles-Chitosan Nanocomposites: A Comparative Study Regarding Different Chemical Syntheses Procedures and Their Antibacterial Effect.: This study explores the synthesis of silver nanoparticles-chitosan nanocomposites using various chemical procedures, including the use of sodium hydroxide, to evaluate their antibacterial properties (Chicea et al., 2024).

  • Using Sparfloxacin-Capped Gold Nanoparticles to Modify a Screen-Printed Carbon Electrode Sensor for Ethanol Determination.: This research demonstrates the modification of sensors using gold nanoparticles with sodium hydroxide, enhancing ethanol detection capabilities (Magesh et al., 2023).

  • Sequence Mining Yields 18 Phloretin C-glycosyltransferases from Plants for the Efficient Biocatalytic Synthesis of Nothofagin and Phloretin-di-C-glycoside.: The study includes the use of sodium hydroxide in biocatalytic processes to synthesize glycosylated compounds efficiently (Putkaradze et al., 2023).

  • Effect of Calcination Temperature and Time on the Synthesis of Iron Oxide Nanoparticles: Green vs. Chemical Method.: This paper investigates the synthesis of iron oxide nanoparticles using sodium hydroxide, comparing green and chemical methods for optimization (Abdullah et al., 2023).

  • Improved Jellyfish Gelatin Quality through Ultrasound-Assisted Salt Removal and an Extraction Process.: Sodium hydroxide is utilized to enhance the extraction process of gelatin from jellyfish, improving its quality through ultrasound-assisted methods (Lueyot et al., 2022).

Analysis Note

Assay (acidimetric,NaOH): ≥ 98.0 %
Carbonate (as Na₂CO₃): ≤ 1.0 %
Identity: passes test
Chloride (Cl): ≤ 0.2 %
Sulfate (SO₄): ≤ 0.05 %
Heavy metals (as Pb): ≤ 0.002 %
Al (Aluminium): ≤ 0.002 %
Fe (Iron): ≤ 0.002 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service