Skip to Content
MilliporeSigma

106331

CellPrime® rAlbumin

recombinant human albumin, EMPROVE® EXPERT

Synonym(s):

Albumin human, rHSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C2936H4624N786O889S41
CAS Number:
Molecular Weight:
66471.53
UNSPSC Code:
12352207
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in Pichia pastoris

Quality Level

sterility

sterile (low bioburden)

product line

EMPROVE® EXPERT

form

liquid

mol wt

monomer ~67 kDa

parameter

2-8 °C temp. range

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

General description

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
recombinant human albumin, expressed in Pichia pastoris

Application

This recombinant variant is recommended for use in absorption, distribution, metabolism, and excretion (ADME) pharmacological research; cell culture; drug delivery research; and cryopreservation of cells.
media manufacturing supplement

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.

Features and Benefits

safe and suitable NAO recombinant supplement

Packaging

  • 1063310020: 20 ml (10%, w/v)
  • 1063310050: 50 ml (10%, w/v)

Shipped in blue ice packs.

Other Notes

Product is manufactured in a facility adhering to Good Manufacturing Practices.
View more information on human serum albumin.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
CellPrime is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

do not freeze


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
106331002004054839606809
106331005004054839608247