Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.05910

Millipore

Maltose monohydrate

suitable for microbiology

Synonym(s):

Maltose monohydrate, 4-O-a-D-Glucopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
MDL number:
UNSPSC Code:
41106212
EC Index Number:
200-716-5
NACRES:
NA.74

pH

4.5-6.0 (20 °C, 50 g/L in H2O)

Quality Level

mp

125 °C (anhydrous)

solubility

470.2 g/L

bulk density

320 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]([C@H](O)[C@@H](O)C=O)[C@H](O)CO.O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

General description

Maltose monohydrate is a sugar with one molecule of water of hydration. It is a white crystalline powder, slightly sweet to taste. Maltose monohydrate is used for the identification of microbes that can ferment it, producing acid and in some cases gas. The identification is carried out using a phenol-red broth base, which contains all the required nutrients for growth of the microorganism except the carbohydrate source which is supplemented by the sugar. If fermentation takes place the indicator (phenol red) turns yellow in color due to the acid produced and if any gas is produced it is collected in the inverted Durham tubes.

Application

Maltose monohydrate is used in microbiology to study the specific nutritional properties of various microorganisms.

Analysis Note

Identity (IR): conforms
Spec. rotation [α²0/D (10 %; water; calc. on anhydrous substance): +135.0 - +139.0 °
Heavy metals (as Pb): ≤ 0.001 %
Water: 4.0 - 7.0 %
Suitability for microbiology: conforms

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service