Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.05853

Supelco

Magnesium nitrate hexahydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Magnesium nitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(NO3)2 · 6 H2O
CAS Number:
Molecular Weight:
256.41
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-826-7
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
420 g/L

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-102.0% (complexometric)

form

solid

potency

5440 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

pH

5.0-7.0 (25 °C, 50 g/L in H2O)

mp

89 °C (for the hexahydrate)

solubility

420 g/L

density

1.46 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.002%

cation traces

As: ≤0.0001%
Ba: ≤0.005%
Ca: ≤0.005%
Fe: ≤0.0005%
K: ≤0.0005%
Mn: ≤0.0005%
NH4+: ≤0.001%
Na: ≤0.0005%
Sr: ≤0.005%
heavy metals: ≤0.0005%

storage temp.

2-30°C

SMILES string

[Mg+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O

InChI

1S/Mg.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

MFUVDXOKPBAHMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Understanding the Roles of Excipients in Moisture Management in Solid Dosage Forms.: This study investigates the roles of various excipients, including magnesium nitrate hexahydrate, in managing moisture in solid dosage forms, highlighting its significance in pharmaceutical formulations (Veronica et al., 2024).

  • Printing Composites with Salt Hydrate Phase Change Materials for Thermal Energy Storage.: This research explores the use of magnesium nitrate hexahydrate as a phase change material in composite printing for thermal energy storage applications, demonstrating its effectiveness in enhancing energy storage efficiency (Lak et al., 2023).

  • How do layered double hydroxides evolve. First in situ insights into their synthesis processes.: The study provides in situ insights into the synthesis processes of layered double hydroxides, utilizing magnesium nitrate hexahydrate as a key reagent in the formation and evolution of these materials (Doungmo et al., 2022).

  • A Wide Bandgap Semiconducting Magnesium Hydrogel: Moisture Harvest, Iodine Sequestration, and Resistive Switching.: This research presents the development of a wide bandgap semiconducting magnesium hydrogel, using magnesium nitrate hexahydrate, with applications in moisture harvesting, iodine sequestration, and resistive switching (Alam et al., 2022).

  • Micro solid phase extraction of parabens from breast milk samples using Mg-Al layered double hydroxide functionalized partially reduced graphene oxide nanocomposite.: This study demonstrates the application of magnesium nitrate hexahydrate in the synthesis of Mg-Al layered double hydroxide for the extraction of parabens from breast milk samples, showcasing its utility in analytical chemistry (Manouchehri et al., 2020).

Linkage

Replaces: MX0060-1

Analysis Note

Assay (complexometric): 99.0 - 102.0 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water, 25 °C): 5.0 - 7.0
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (ACS): ≤ 0.0005 %
As (Arsenic): ≤ 0.0001 %
Ba (Barium): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mn (Manganese): ≤ 0.0005 %
Na (Sodium): ≤ 0.0005 %
NH₄ (Ammonium): ≤ 0.001 %
Sr (Strontium): ≤ 0.005 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service