Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.05591

Supelco

Sodium hydroxide solution

min. 27% (1.30), for analysis (for the determination of nitrogen) EMSURE®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

product name

Sodium hydroxide solution min. 27% (1.30), for analysis (for the determination of nitrogen) EMSURE®

vapor pressure

3 mmHg ( 37 °C)

Quality Level

product line

EMSURE®

form

liquid

concentration

≥27.0% (acidimetric)

impurities

≤0.0001% Total nitrogen (N)
≤0.0005% Al (Aluminium)
≤0.0005% Fe (Iron)
≤0.0005% Heavy metals (as Pb)
≤0.0005% Phosphate (PO4)
≤0.001% Silicate (SiO2)
≤0.003% Sulfate (SO4)
≤0.004% Chloride (Cl)
≤1.0% Carbonate (as Na2CO3)

pH

14 (20 °C in H2O)

density

1.30 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric): ≥ 27.0 %
Carbonate (as Na₂CO₃): ≤ 1.0 %
Chloride (Cl): ≤ 0.004 %
Phosphate (PO₄): ≤ 0.0005 %
Silicate (SiO₂): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.003 %
Total nitrogen (N): ≤ 0.0001 %
Heavy metals (as Pb): ≤ 0.0005 %
Al (Aluminium): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service