Skip to Content
MilliporeSigma

1.05151

Potassium sulfate

powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, E 515

Synonym(s):

Potassium sulfate, Potassium pyrosulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
K2O4S
CAS Number:
Molecular Weight:
174.26
UNSPSC Code:
12352302
E Number:
E515
EC Index Number:
231-915-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

agency

BP, JP, Ph. Eur.

product line

EMPROVE® ESSENTIAL

form

fine powder, powder

pH

5.5-8.5 (20 °C, 50 g/L in H2O)

bp

1689 °C/1013 hPa

mp

1069 °C

solubility

111 g/L

density

2.66 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[K+].[K+].[S](=O)(=O)([O-])[O-]

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium sulfate powder Emprove® Essential is mainly used in highly regulated pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Potassium sulfate


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
105151100004022536061641
105151902904022536946146
105151999904022536550091