Skip to Content
MilliporeSigma

1.05134

Potassium sulfide

small lumps for analysis EMSURE®

Synonym(s):

Potassium sulfide, Potassium monosulfide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(l · K2S)(m · K2Sx)(n · K2S2O3)
CAS Number:
NACRES:
NA.21
UNSPSC Code:
12352302
MDL number:
Grade:
reagent grade
Form:
solid
Solubility:
500 g/L
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

reagent grade

product line

EMSURE®

form

solid

concentration

≥44% (complexometric K2S basis)

pH

13 (20 °C, 10 g/L in H2O)

mp

200-250 °C

solubility

500 g/L

density

1.65 g/cm3 at 20 °C

bulk density

1000‑1200 kg/m3

anion traces

sulfite (SO32-): ≤1%, thiosulfate (S2O32-): ≤20

cation traces

N: ≤0.01%

storage temp.

no temp limit

InChI

1S/4K.H2O3S2.S/c;;;;1-5(2,3)4;/h;;;;(H2,1,2,3,4);/q;;2*+1;;/p-2

InChI key

PFSBHPPVZHPSCB-UHFFFAOYSA-L

Analysis Note

Assay (complexometric, as K₂S): ≥ 44 %
Sulfite (SO₃): ≤ 1 %
Thiosulfate (S₂O₃): about 20
Total nitrogen (N): ≤ 0.01 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

supp_hazards

wgk

WGK 3

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library