Skip to Content
MilliporeSigma

1.05099

Potassium phosphate dibasic trihydrate

for analysis EMSURE®

Synonym(s):

di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
HK2O4P · 3H2O
CAS Number:
Molecular Weight:
228.22
UNSPSC Code:
12352302
EC Index Number:
231-834-5
NACRES:
NA.21
MDL number:
Assay:
≥99.0% (acidimetric)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

di-Potassium hydrogen phosphate trihydrate, for analysis EMSURE®

Quality Segment

product line

EMSURE®

assay

≥99.0% (acidimetric)

form

solid

pH

9.2-9.4 (20 °C, 50 g/L in H2O)

mp

340 °C (anhydrous substance; decomposes)

bulk density

800 kg/m3

anion traces

chloride (Cl-): ≤0.001%, sulfate (SO42-): ≤0.005%

cation traces

As: ≤0.00005%, Fe: ≤0.0005%, N: ≤0.001%, Na: ≤0.1%, heavy metals (as Pb): ≤0.001%

storage temp.

15-25°C

SMILES string

[K+].[K+].[P](=O)([O-])([O-])O.O.O.O

InChI

1S/2K.H3O4P.3H2O/c;;1-5(2,3)4;;;/h;;(H3,1,2,3,4);3*1H2/q2*+1;;;;/p-2

InChI key

XQGPKZUNMMFTAL-UHFFFAOYSA-L

Analysis Note

Assay (acidimetric): ≥ 99.0 %
pH-value (5 %; water): 9.1 - 9.3
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.00005 %
Fe (Iron): ≤ 0.0005 %
Na (Sodium): ≤ 0.1 %

Other Notes

Replaces: PX1571; PX1571-1; PX1571-20

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library