Skip to Content
MilliporeSigma

1.04852

Potassium hydrogen carbonate

EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, FCC, E 501

Synonym(s):

Potassium hydrogen carbonate, Potassium bicarbonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
UNSPSC Code:
12352302
E Number:
E501
EC Index Number:
206-059-0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, Ph. Eur., USP

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

solid

pH

8.4-8.6 (20 °C, 99 g/L in H2O)

solubility

362 g/L

density

2.17 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[K+].[O-]C(=O)[O-].[H+]

InChI

1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

TYJJADVDDVDEDZ-UHFFFAOYSA-M

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium hydrogen carbonate (synonym potassium bicarbonate) is a commonly used ingredient in both food and pharmaceutics. In pharmaceutical applications, potassium hydrogen carbonate is primarily used as a source of carbon dioxide in oral formulations such as effervescent tablets or granules.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Potassium hydrogen carbonate


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library