Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04728

Millipore

myo-Inositol

suitable for microbiology

Synonym(s):

myo-Inositol, Hexahydroxycyclohexane, Cyclohexanehexol, meso-Inositol, Myo-Inosit

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
41106212
EC Index Number:
201-781-2
NACRES:
NA.74

Assay

≥99% (HPLC)

Quality Level

form

solid

pH

5-7 (20 °C, 100 g/L in H2O)

mp

225-227 °C

solubility

143 g/L

application(s)

microbiology

storage temp.

15-25°C

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H

InChI key

CDAISMWEOUEBRE-UHFFFAOYSA-N

Analysis Note

Assay (HPLC): ≥ 99 %
Identity (IR): conforms
Melting range (lower value): ≥ 224 °C
Melting range (upper value): ≤ 227 °C
Heavy metals (as Pb): ≤ 0.001 %
Water: ≤ 0.5 %
Suitability for microbiology: conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service