Skip to Content
MilliporeSigma

1.04445

Benzonase® Salt Tolerant Endonuclease

greener alternative

EMPROVE® EXPERT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

EMPROVE® EXPERT

Quality Segment

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

pharma/biopharma processes

greener alternative category

storage temp.

-10 to -25°C

General description

The best-in-class solution for nucleic acid removal at high salt concentration in Bioprocessing
We are committed to bringing you Greener AlternativeProducts, which belongs to one of the four categories of greener alternatives .This product is a Design for Sustainability (DfS) developed product. Click hereto view its DfS scorecard.

Application

The Benzonase® Salt Tolerant endonuclease Emprove® Expert is a new enzyme, different in amino acids sequence from other Benzonase® endonuclease references. It was designed with state-of-the-art protein engineering capabilities, to ensure the highest activity at high salt concentrations. The protein is a monomer with a molecular weight of about 27 kDa . The protein has an isoelectric point (pI) at pH 9.68. It is functional between pH 6 and 10, and from 0 °C to above 37 °C. Mg2+ (1–2 mM) is required for enzyme activity.

Packaging

  • 80,000 Units
  • 400,000 Units
  • 4,000,000 Units + 40, 000 Units tailgate sample

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 1



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library